Anti-LST-3TM12 Rabbit Polyclonal Antibody
AVIVARP44578_P050
- Antibody type:Primary
- Antigen name:solute carrier organic anion transporter family, member 1B7 (non-functional)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:338821
- Antigen synonyms:SLC21A21|LST-3|LST3|LST-3TM12
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: LST-3TM12
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human