Anti-ATP2B3 Rabbit Polyclonal Antibody
Catalog # AVIVARP44390_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:ATPase, Ca++ transporting, plasma membrane 3
- Antigen symbol:ATP2B3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig,Human,Bovine,Guinea pig,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:492
- Antigen synonyms:OPCA|CFAP39|SCAX1|CLA2|PMCA3a|PMCA3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ATP2B3 (ATPase, Ca++ transporting, plasma membrane 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rat