Order Entry
Ireland
ContactUsLinkComponent
Anti-SLC9C1 Rabbit Polyclonal Antibody
Anti-SLC9C1 Rabbit Polyclonal Antibody
Catalog # AVIVARP44166_P050
undefined
Anti-SLC9C1 Rabbit Polyclonal Antibody
Catalog # AVIVARP44166_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 9, subfamily C (Na+-transporting carboxylic acid decarboxylase), member 1
  • Antigen symbol:
    SLC9C1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    288117
  • Antigen synonyms:
    Slc9a10|Gm610
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: SLC9C1 (solute carrier family 9, subfamily C (Na+-transporting carboxylic acid decarboxylase), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: