Order Entry
Ireland
ContactUsLinkComponent
 
Anti-FAM46A Rabbit Polyclonal Antibody
  AVIVARP42484_P050
Anti-FAM46A Rabbit Polyclonal Antibody
  AVIVARP42484_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    Family with sequence similarity 46, member A
  • Antigen symbol:
    FAM46A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    55603
  • Antigen synonyms:
    XTP11|C6orf37
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: FAM46A (family with sequence similarity 46, member A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: