Anti-TRMT10A Rabbit Polyclonal Antibody
Catalog # AVIVARP41144_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:tRNA methyltransferase 10A
- Antigen symbol:TRMT10A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:93587
- Antigen synonyms:TRM10|HEL-S-88|RG9MTD2|MSSGM
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TRMT10A (tRNA methyltransferase 10 homolog A (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: