Anti-SRPR Rabbit Polyclonal Antibody
Catalog # AVIVARP40460_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Signal recognition particle receptor
- Antigen symbol:SRPR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Bovine,Guinea pig,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:6734
- Antigen synonyms:Sralpha|DP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SRPR (signal recognition particle receptor (docking protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Rat