Anti-ZUFSP Rabbit Polyclonal Antibody
Catalog # AVIVARP39842_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Zinc finger with UFM1-specific peptidase domain
- Antigen symbol:ZUFSP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:221302
- Antigen synonyms:dJ412I7.3|C6orf113
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ZUFSP (zinc finger with UFM1-specific peptidase domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat