Order Entry
Ireland
ContactUsLinkComponent
 
Anti-SNAPC1 Rabbit Polyclonal Antibody
  AVIVARP38227_T100
undefined
Anti-SNAPC1 Rabbit Polyclonal Antibody
  AVIVARP38227_T100

 

  • Antibody type:
    Primary
  • Antigen name:
    small nuclear RNA activating complex, polypeptide 1, 43kDa
  • Antigen symbol:
    SNAPC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    6617
  • Antigen synonyms:
    PTFgamma|SNAP43
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: SNAPC1 (small nuclear RNA activating complex, polypeptide 1, 43kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human