Anti-DRGX Rabbit Polyclonal Antibody
Catalog # AVIVARP36803_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Dorsal root ganglia homeobox
- Antigen symbol:DRGX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:252880
- Antigen synonyms:Drg11|Prrxl1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: DRGX (dorsal root ganglia homeobox)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat