Anti-HOXA7 Rabbit Polyclonal Antibody
AVIVARP32637_P050
- Antibody type:Primary
- Antigen name:homeobox A7
- Antigen symbol:HOXA7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:3204
- Antigen synonyms:HOX1|HOX1.1|HOX1A|ANTP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: HOXA7 (homeobox A7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: