Anti-GTF2H4 Rabbit Polyclonal Antibody
AVIVARP32598_P050
- Antibody type:Primary
- Antigen name:General transcription factor 2H, polypeptide 4, 52kDa
- Antigen symbol:GTF2H4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2968
- Antigen synonyms:TFB2|TFIIH|P52
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: GTF2H4 (general transcription factor IIH, polypeptide 4, 52kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: