Anti-CCRN4L Rabbit Polyclonal Antibody
Catalog # AVIVARP32536_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Nocturnin
- Antigen symbol:NOCT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:25819
- Antigen synonyms:CCR4L|Ccr4c|CCRN4L|NOC
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CCRN4L (CCR4 carbon catabolite repression 4-like (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: