Anti-ZNF541 Rabbit Polyclonal Antibody
AVIVARP30068_T100
- Antibody type:Primary
- Antigen name:zinc finger protein 541
- Antigen symbol:ZNF541
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:84215
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: ZNF541 (zinc finger protein 541)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: