Anti-F2R Rabbit Polyclonal Antibody
Catalog # BOSSBS-0828R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Coagulation Factor 2 Receptor
- Antigen symbol:F2R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:PAR1|Thrombin Receptor|PAR-1|HTR|CF2R|TR
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: F2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat