Order Entry
Ireland
ContactUsLinkComponent
Hydrochloric acid 17%, EMPROVE® EXPERT for biopharmaceutical production
Hydrochloric acid 17%, EMPROVE® EXPERT for biopharmaceutical production
Catalog # 4.80708.9950
CAS Number:  
Hydrochloric acid 17%, EMPROVE® EXPERT for biopharmaceutical production
Catalog # 4.80708.9950
CAS Number:  

Specifications

  • Pk:
    950 L
  • Seq:
    5783

Specifications

About this item

 :
ADR:8,II
 :36.46 g/mol
EINECS:231-595-7
UN:1789
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324