Order Entry
Ireland
ContactUsLinkComponent
Inoculating loops, stainless steel
Inoculating loops, stainless steel
Catalog # WART6.000.373
Supplier:  WARTEWIG
undefined
Inoculating loops, stainless steel
Catalog # WART6.000.373
Supplier:  WARTEWIG

Specifications

  • Ø:
    3 mm
  • Description:
    Inoculation loop, stainless steel wire Ø 0,6 mm
  • Seq:
    02
  • Pk:
    10

Specifications

About this item