Order Entry
Ireland
ContactUsLinkComponent
Sleeve protector, cut resistant, HyFlex® 11-200
Sleeve protector, cut resistant, HyFlex® 11-200
Catalog # ANSE11-200
Supplier:  Ansell
undefined
Sleeve protector, cut resistant, HyFlex® 11-200
Catalog # ANSE11-200
Supplier:  Ansell
Specifications
  • Garment application:
    Industrial,
    Mechanical protection
  • Gender:
    Unisex
  • Risk area:
    Cuts,
    HiVis
  • Colour:
    Hi-vis yellow
  • Length:
    475 mm
  • Fabric:
    Polyester/nylon/glass fibre
  • Sleeve or leg length:
    Long
  • Pk:
    12
Specifications
About this item
This high visibility sleeve with Level 5 cut performance and Level 1 thermal protection is knitted from polyester, nylon and glass fibres. The anatomic design with adjustable forearm area enables freedom of movement so workers can do the job without constraints, increasing productivity.
  • Most comfortable sleeve in the Ansell range: Generous space in the arm area reduces sweat and itchiness Better abrasion resistance (Level 2) and washability mean HyFlex® 11-200 offers improved total cost of ownership Due to its ergonomic design and fixing system, worker acceptance will increase, leading to fewer accidents and injuries
Ideal for use in body shops (excluding welding), for glass and metal sheet handling and for stamping operations.
Gauge: 13
Certifications: EN 388 (2542), EN 407 (X1XXXX); Cat. II