Order Entry
Ireland
ContactUsLinkComponent
Triethylamine ≥99.8%, HiPerSolv CHROMANORM® for LC-MS
Flammable
Irritant
Corrosive
Triethylamine ≥99.8%, HiPerSolv CHROMANORM® for LC-MS
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Irritant
Corrosive
Triethylamine ≥99.8%, HiPerSolv CHROMANORM® for LC-MS
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.8 %
  • Evaporation residue:
    Max. 0.0001 %
  • Water:
    Max. 0.02 %
  • Transmittance (230 nm) (0.1 %):
    Min. 10 %
  • Transmittance (240 nm) (0.1 %):
    Min. 50 %
  • Transmittance (245 nm) (0.1 %):
    Min. 80 %
  • Transmittance (250 nm) (0.1 %):
    Min. 95 %
  • Transmittance (255 nm) (0.1 %):
    Min. 99 %
  • Suitable for LC-MS (0.1 %):
    Passes test

Specification Test Results

Frequently Bought Together