Order Entry
Ireland
ContactUsLinkComponent
Visual colour kits, freshwater, CA-10WR
Visual colour kits, freshwater, CA-10WR
Catalog # HACH143801
Supplier:  Hach
CAS Number:  
undefined
Visual colour kits, freshwater, CA-10WR
Catalog # HACH143801
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    CA-10WR test kit for freshwater including carbon dioxide, dissolved oxygen, and pH test kits
Specifications
About this item
Uses simple, fast colour disc and drop count methods to measure carbon dioxide, dissolved oxygen, and pH for applications in water ecology
Delivery: Kit includes seven reagents, clippers, bottles, sample cell with cap, colour disc, viewing tubes, comparator box, measuring tube, instructions and carrying case.