Polyvinylpyrrolidone, Divergan® RS, Supelco®
Catalog # 1.07302.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Identity (IR spectrum):conforms
- Molar Mass:40000 g/mol
- Bulk Density:330 kg/m3
- Density:1.2 g/cm3 (20 °C)
- Ignition Point:440 °C
Specification Test Results
About this item
:
Melting Pt:220 °C (dec.)
Density:1,2 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
CAS Number:9003-39-8
Formula:(C₆H₉NO)n
MDL Number:MFCD01076626