Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Syringe Needles
Replacement needles for gold standard autosampler syringes
Replacement needles for gold standard autosampler syringes
Replacement needles for gold standard autosampler syringes
Catalog # HEWL5181-3319
Supplier:
Agilent
Expand image
Replacement needles for gold standard autosampler syringes
Catalog # HEWL5181-3319
Supplier:
Agilent
Specifications
Description:
Replacement needle, 23S - 26S gauge, for 10 µl Gold standard autosampler syringes
Pk:
3
Specifications
About this item
Product Details
Product Details