Scoops
Supplier: Adelphi Pharma Hygiene Products
Jump to:
About this item
Stainless steel 316L.
- Heavy-duty
- Crevice- and pit-free
- Polished throughout
- With round handle
2 Options Available Below
Product Details & Documents
Stainless steel 316L.
- Heavy-duty
- Crevice- and pit-free
- Polished throughout
- With round handle
Product Family Options
Product Information