Sodium hydrogen sulphate monohydrate 35,0 - 36,5% (by acidimetry), crystals, BAKER ANALYZED®, J.T.Baker®
Catalog # 3534-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:35.0 - 36.5 %
- Assay (H₂SO₄) (by acidimetry):35.0 - 36.5 %
- Insoluble Matter and NH₄OH Precipitate:≤0.005 %
- Chloride (Cl):≤0.001 %
- Phosphate (PO₄):≤0.001 %
- Calcium and Magnesium (as Ca) (by EDTA titration):≤0.005 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Trace Impurities - Arsenic (As):≤1 ppm
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
Specification Test Results
About this item
Synonyms: Sodium bisulphate monohydrate
:
:138.08 g/mol
Merck Index:12,08729
Melting Pt:183 °C
Density:1,8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-665-7
UN:3260
ADR:8,III
:GHS05
CAS Number:10034-88-5
Formula:NaHSO₄·H₂O
MDL Number:MFCD00149210