Florisil® 60 - 100 mesh, J.T.Baker®
Catalog # M368-07
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 10 %
- Mesh - On U.S. No. 60 Sieve:≤10 %
- Mesh - Thru U.S. No. 100 Sieve:≤10 %
Specification Test Results
About this item
:
:100.38 g/mol
Density:2,51 Kg/L
EINECS:215-681-1
CAS Number:1343-88-0
Formula:MgO₃Si
MDL Number:MFCD00078249