Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrodes
pH Electrodes
Accessories for pH combination electrodes, AquaLine
Accessories for pH combination electrodes, AquaLine
Accessories for pH combination electrodes, AquaLine
Catalog # 662-6191
Supplier:
SI Analytics
Expand image
Accessories for pH combination electrodes, AquaLine
Catalog # 662-6191
Supplier:
SI Analytics
Specifications
Description:
Tapper adapter, NS 14,5, PTFE, for electrodes with Ø 12 mm
Pk:
1
Specifications
About this item
Product Details
Product Details