Order Entry
Ireland
ContactUsLinkComponent
L(+)-Ascorbic acid ≥99.5%, powder, BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
L(+)-Ascorbic acid ≥99.5%, powder, BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
Catalog # B581-05
Supplier:  Avantor
CAS Number:  
undefined
L(+)-Ascorbic acid ≥99.5%, powder, BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
Catalog # B581-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.5 %
  • Assay (C₆H₈O₆):
    ≥ 99.5 %
  • Specific Rotation,α²⁵D(c=10 in water)(+):
    20.5 - 21.5 Degree
  • Residue after Ignition:
    ≤ 0.10 %
  • Loss on Drying at 105°C:
    ≤ 0.1 %
  • Heavy Metals (as Pb):
    ≤ 0.001 %
  • Trace Impurities - Iron (Fe):
    ≤ 0.001 %

Specification Test Results

About this item

 :
 :176.13 g/mol
Merck Index:13,00837
Boiling Pt:553 °C (1013 hPa)
Melting Pt:190…192 °C
Density:1,694 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-066-2
REACH:exempt
CAS Number:50-81-7
Formula:C₆H₈O₆
MDL Number:MFCD00064328