Order Entry
Ireland
ContactUsLinkComponent
Counting chamber, McMaster
Counting chamber, McMaster
Catalog # HAWKZ11000
Supplier:  HAWKSLEY AND SONS
Counting chamber, McMaster
Catalog # HAWKZ11000
Supplier:  HAWKSLEY AND SONS
Specifications
  • Depth of chamber:
    1,5 mm
  • Type:
    McMaster
  • Description:
    Counting chamber
  • Pk:
    1
Specifications
About this item
Counting chamber with cover glass and two counting areas.
  • 10×10 mm grid etched on the under side of the top cover
Suitable for worm egg counts.