Order Entry
Ireland
ContactUsLinkComponent
PCR racks
PCR racks
  DIUKRTP/78135-N
PCR racks
  DIUKRTP/78135-N
  • Colour:
    Natural
  • Description:
    PCR rack with lid,PP/PS
  • Pk:
    5
PP, with lid.
  • A range of racks that will hold either strips of 8 or 12×0,2 ml tubes
  • Racks have alphanumeric grid for sample and well identification
  • Sliding lid for added security and when lid is closed the racks are stackable
  • Each rack has label/tag holder
  • Can be used by attaching multi-channel pipette reservoir