Order Entry
Ireland
ContactUsLinkComponent
Cleanroom gloves, latex, Multi-Flex® Clean Class 100
Cleanroom gloves, latex, Multi-Flex® Clean Class 100
Supplier:  CARDINAL HEALTH
Certificates
Cleanroom gloves, latex, Multi-Flex® Clean Class 100
Supplier:  CARDINAL HEALTH
Specifications
  • Catalog No:
  • CARD2Y1607
  • CARD2Y1606
  • CARD2Y1604
  • CARD2Y1602
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5