Order Entry
Ireland
ContactUsLinkComponent
SPE plates, 96-well, HyperSep™, SAX
SPE plates, 96-well, HyperSep™, SAX
  HYPE60300-567
SPE plates, 96-well, HyperSep™, SAX
  HYPE60300-567
  • Description:
    96-well plate, HyperSep™ SAX
  • Seq:
    07
  • Filling:
    50 mg
  • Pk:
    1
Thermo Scientific™ HyperSep SAX Strong Anion Exchanger SPE 96-Well Plates and Individual Wells feature a strong anion exchange sorbent for extraction of weak acids. The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.