Water purification systems, AFS® 8/16/24
Supplier: Merck
Certificates
About this item
Economical water system, producing CLRW (Clinical Laboratory Reagent Water) quality with a dispensing flow rate to analyser up to 2 l/min. For clinical analysers with pure water needs up to 480 L daily
- Cost-effective: Optimised lifetimes for pretreatment and polishing packs, low feed water use
- Full monitoring and automatic water quality archiving; information can be stored for up to 6 months
- Small footprint, allowing installation wherever it’s convenient: on the wall, on or under the bench, or on a cart
- Intuitive icon-based system display facilitates maintenance
- Ergonomic pack locking system makes pack changes easy
Complementary water purification techniques, including complete pretreatment, advanced reverse osmosis (RO), exposure to a bactericidal UV lamp, and final 0,22 μm filtration, provide consistent water quality meeting CLRW standards.
Specifications
- Catalog No:
- MILLZAFS0080UK
- MILLZAFS0160UK
- MILLZAFS0240UK
- Model:
- AFS® 8
- AFS® 16
- AFS® 24
- Feed water:
- Portable tap water at a maximum pressure of 6 bar
- Portable tap water at a maximum pressure of 6 bar
- Portable tap water at a maximum pressure of 6 bar
- Production flow rate:
- 8 L/h @ 25 °C
- 16 L/h @ 25 °C
- 24 L/h @ 25 °C
- TOC content:
- <30 ppb
- <30 ppb
- <30 ppb
- Resistivity at +25 °C:
- >15 MΏ.cm
- >15 MΏ.cm
- >15 MΏ.cm
- Bacteria:
- < 1 CFU/ml
- < 1 CFU/ml
- < 1 CFU/ml
- W×D×H:
- 268×426×585 mm
- 268×426×585 mm
- 268×426×585 mm
- Weight:
- <28 kg
- <28 kg
- <28 kg
Specifications
Frequently Bought Together