Neutralising liquids, Spilfyter® Kolorsafe
Catalog # NPSD410001
Supplier: NPS
Specifications
- Description:Neutralising liquid, Spilfyter® Kolorsafe for acids
- Packed:0,95 litre bottles
- Pk:12
Specifications
About this item
Neutralise hazardous caustic and acid spills (including hydrofluoric acid) with these easy-to-use liquid neutralisers.
- Liquid formula generates less gas and heat during neutralisation than dry equivalent products
- For treating spills on vertical or sloping surfaces
- Kolorsafe color change process which indicates when enough neutraliser has been applied to the spill and spill is safe for absorption and disposal