Cleanroom overalls, Primeguard Plus
These overalls are ideal for use in cleanrooms and laboratories where the microporous laminate provides strength with excellent comfort. Repellent to water based liquids and low hazard chemicals.
- Very soft texture ensuring high comfort for extended periods
- Good water vapour transmission rate and excellent blood barrier
- With hood
Colour: White
- Catalog No:
- VEROPG6L30
- VEROPG6X30
- Model:
- Primeguard Plus
- Primeguard Plus
- Garment application:
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
Frequently Bought Together