Vertical electrophoresis systems, omniPAGE mini
Supplier: CLEAVER SCIENTIFIC
Certificates
About this item
By combining both functionality and ease of use, the OmniPAGE CVS10 systems provide complete flexibility for individual research needs. They can accommodate up to 4 handcast gels and 2 commercial precast gels and the sliding clamp technology within the PAGE insert facilitates fast, intuitive leak-free casting.
- Run a maximum of 4 gels within an hour
- Perform 2-D and blotting within a day
- Undertake discovery projects
- Screen new samples and evaluate sample preparation conditions
Delivery: Each complete system includes glass plates with bonded 1,0 mm thick spacers, 2×12 sample combs, cooling pack and dummy plate.
Specifications
- Catalog No:
- CLEVVS10DSYS
- CLEVVS10D
- Model:
- omniPAGE mini
- omniPAGE mini
- Gel size W×H:
- 75×80 mm
- 75×80 mm
- Buffer volume, gel tank:
- 250-1200 ml
- 250-1200 ml
- Glass plate size (W×H):
- 100×100 mm
- 100×100 mm
- Unit W×H×D:
- 190×150×130 mm
- 190×150×130 mm
Specifications
Frequently Bought Together