Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Notebooks
VWR®, Cleanroom Spiral Notebooks
VWR®, Cleanroom Spiral Notebooks
VWR®, Cleanroom Spiral Notebooks
Catalog # VWRU51280-888
Supplier:
VWR Collection
Expand image
VWR®, Cleanroom Spiral Notebooks
Catalog # VWRU51280-888
Supplier:
VWR Collection
Specifications
Paper size:
216×140 mm
Type:
Ruled
Colour:
Grey
Description:
Cleanroom spiral notebook, Critical print™
Seq:
01
Pk:
20
Specifications
About this item
Product Details
Product Details