Foam head swabs, TechSwabs™
Supplier: INTERTRONICS
Certificates
About this item
The foam head swabs are manufactured with 100 pores per inch reticulated polyurethane foam.
- Flexible tips offer versatility and softer touch while rigid heads allow for more aggressive cleaning
- Heat seal to prevent tip loss even while saturated with solvent
- Excellent absorbency with reduced contamination
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together