Order Entry
Ireland
ContactUsLinkComponent
Enzymatic digester, GDE
Enzymatic digester, GDE
Supplier:  VELP SCIENTIFIC
Certificates
undefined
Enzymatic digester, GDE
Supplier:  VELP SCIENTIFIC
Specifications
  • Catalog No:
  • VELPSA30400209
  • Temperature range:
  • RT*...105 °C
  • Heating capacity:
  • 900 W
  • Ext. W×D×H:
  • 413×410×295 mm
  • Weight:
  • 6,2 kg