Order Entry
Ireland
ContactUsLinkComponent
Nervous system relief model
Nervous system relief model
Catalog # BINH1000231
Supplier:  3B Scientific
undefined
Nervous system relief model
Catalog # BINH1000231
Supplier:  3B Scientific

Specifications

  • Description:
    Nervous system relief model
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

This nervous system relief model shows a schematic representation of the central and peripheral nervous system. An excellent nervous system model to study the structure of the human nervous system. This nervous system model is ½ life size and is a great teaching tool.

  • Delivered on baseboard

Dimensions: 80×33×6 cm

Weight: 3,5 kg