Order Entry
Ireland
ContactUsLinkComponent
Alcohol sterile, blended with WFI, Klercide 60/40
Alcohol sterile, blended with WFI, Klercide 60/40
Catalog # SHIE3054760
Alcohol sterile, blended with WFI, Klercide 60/40
Catalog # SHIE3054760
Specifications
  • Description:
    Sterile alcohol blended with WFI,capped
  • Environmentally Preferable:
  • Packed:
    5 litre
  • Pk:
    4
Specifications
About this item
60% alcohol blended with Water for Injection. Supplied sterile by gamma irradiation and double bagged.
  • Endotoxin levels guaranteed to be below 0,25 EU/ml
  • Ideal for Grade A and B cleanrooms and product contact areas
  • SteriShield Delivery System™ ensures 6 months in-use shelf life
  • Certificates of Analysis, Irradiation and Sterility with every batch
  • Fully validated and CEN tested
Certifications: Bactericidal EN 1276; Fungicidal EN 1650; Bactericidal and Fungicidal EN 13697;