Order Entry
Ireland
ContactUsLinkComponent
Cut resistant gloves, uvex C500 foam
Cut resistant gloves, uvex C500 foam
Supplier:  UVEX
Certificates
undefined
Cut resistant gloves, uvex C500 foam
Supplier:  UVEX
Specifications
  • Catalog No:
  • 112-4498
  • 112-4494
  • 112-4495
  • 112-4496
  • 112-4497
  • Model:
  • foam
  • foam
  • foam
  • foam
  • foam
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Viscose/Dyneema®/glass/PA
  • Viscose/Dyneema®/glass/PA
  • Viscose/Dyneema®/glass/PA
  • Viscose/Dyneema®/glass/PA
  • Viscose/Dyneema®/glass/PA
  • Finish:
  • High Performance Elastomer (HPE) SoftGrip foam coating on palm and fingertips
  • High Performance Elastomer (HPE) SoftGrip foam coating on palm and fingertips
  • High Performance Elastomer (HPE) SoftGrip foam coating on palm and fingertips
  • High Performance Elastomer (HPE) SoftGrip foam coating on palm and fingertips
  • High Performance Elastomer (HPE) SoftGrip foam coating on palm and fingertips
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4X42C)
  • EN 388:2016 (4X42C)
  • EN 388:2016 (4X42C)
  • EN 388:2016 (4X42C)
  • EN 388:2016 (4X42C)