Order Entry
Ireland
ContactUsLinkComponent
Barrier tapes, suspected carcinogen hazard
Barrier tapes, suspected carcinogen hazard
Catalog # LIMP661-029
Supplier:  PREMIER LIMPET
Barrier tapes, suspected carcinogen hazard
Catalog # LIMP661-029
Supplier:  PREMIER LIMPET

Specifications

  • Width:
    25,0 mm
  • Description:
    Suspected carcinogen warning tape
  • Environmentally Preferable:
  • Pk:
    1 Roll

Specifications

About this item

These barrier tapes are made of vinyl.

Used for packaging, streamering across cabinets and for general use as signs

Length: 66,0 m

Colour: Black on yellow