Disposable particle filtering respirators, Comfort-Line, zetMask®
Catalog # ZVGA30941-94
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Specifications
- Type:Red
- Seq:01
- Pk:20
- With valve:-
- Class:FFP2
Specifications
About this item
The particle-filtering half-masks of the Comfort line feature an extremely low breathing resistance combined with a high filtering capacity. This also applies for masks without exhalation valve.
- High quality mask
- Flat folding , individually hygienically packed
- With elastic bands
- Colour coded
Certifications: EN 149:2001