Tip cleaning sponge, Hexacon
CAULSP8141C
- Description:Sponge with one hole, compressed
- L×W:57×89 mm
- Pk:10
This cellulose sponge with one hole provides an effective method to clean soldering residues from a hot tip. With regular use additional abrasive cleaning can become unnecessary.
- Purpose-made for tip cleaning
- Large continuous surface area
- Very low sulphur and acid content prevents tip contamination
- Fine pore texture suits all tip sizes