Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boxes
Storage Boxes
Spare clips for Gastronorm container
Spare clips for Gastronorm container
Spare clips for Gastronorm container
Catalog # 216-1469
Supplier:
Azpack
Expand image
Spare clips for Gastronorm container
Catalog # 216-1469
Supplier:
Azpack
Specifications
Description:
Pack with each two brown, two green, two yellow, two white, two red and two blue Gastronorm spare clips
Pk:
12
Specifications
About this item
Product Details
Product Details