Jars, wide mouth
Supplier: SEMADENI
Certificates
About this item
Jars, made of HDPE or PP, translucent. Resistant to shape, solid design, optionally with close lid.
- Good chemical resistance
- Good temperature resistance
- Lightweight
- Lids optionally available
Very cost-effective, also suitable as a disposable cup or packaging.
Frequently Bought Together