Calcein AM
About this item
The fluorescent dye calcein acetoxymethylester (calcein AM) can be used to label cells when performing analyses such as tumour cell invasion, endothelial cell migration, endothelial cell tubulogenesis, and other cell-based assays.
- Non fluorescent cell viability indicator
- Cell permeant compound that is hydrolysed by intracellular esterases in the fluorescent anion calcein
Calcein AM can be used to fluorescently pre- and post-label viable cells and perform kinetic and endpoint experiments, respectively.
The fluorescent dyes, calcein acetoxymethylester (calcein AM), can be used with Falcon* Corning FluoroBlok* cell culture inserts and insert systems to label cells when performing analyses such as tumor cell invasion, endothelial cell migration, and other cell-based assays
Caution: NOTE: Cells are known to tolerate different fluorescent dyes for varying amounts of time in culture. Cells labelled with calcein AM should be exposed to the dye for less than eight hours.
- CAS Number:148504-34-1
- Formula:C₄₆H₄₆N₂O₂₃
- MDL Number:MFCD05861516
- Molecular Weight:994.86 g/mol
2 Options Available Below
Product Details & Documents
The fluorescent dye calcein acetoxymethylester (calcein AM) can be used to label cells when performing analyses such as tumour cell invasion, endothelial cell migration, endothelial cell tubulogenesis, and other cell-based assays.
- Non fluorescent cell viability indicator
- Cell permeant compound that is hydrolysed by intracellular esterases in the fluorescent anion calcein
Calcein AM can be used to fluorescently pre- and post-label viable cells and perform kinetic and endpoint experiments, respectively.
The fluorescent dyes, calcein acetoxymethylester (calcein AM), can be used with Falcon* Corning FluoroBlok* cell culture inserts and insert systems to label cells when performing analyses such as tumor cell invasion, endothelial cell migration, and other cell-based assays
Caution: NOTE: Cells are known to tolerate different fluorescent dyes for varying amounts of time in culture. Cells labelled with calcein AM should be exposed to the dye for less than eight hours.
- CAS Number:148504-34-1
- Formula:C₄₆H₄₆N₂O₂₃
- MDL Number:MFCD05861516
- Molecular Weight:994.86 g/mol
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



