Order Entry
Ireland
ContactUsLinkComponent
Cleanroom wipes for disinfectants, CoverMAX®
Cleanroom wipes for disinfectants, CoverMAX®
Catalog # FOAMFP4290ST
Supplier:  FOAMTEC
undefined
Cleanroom wipes for disinfectants, CoverMAX®
Catalog # FOAMFP4290ST
Supplier:  FOAMTEC
Specifications
  • L×W:
    228×228 mm
  • Pk:
    100
Specifications
About this item
This triple-layered wipe is designed to aid decontamination procedures in Pharmaceutical and Medical Device manufacturing facilities.
  • Can hold 8 to 10× the volume of fluid compared to other wipes allowing for much quicker application of cleaning fluids to large areas such as glass windows, stainless steel tanks and piping
  • Ideal for applying disinfectants to difficult to clean surfaces such as conveyors
  • Resists shedding abrasion generated fibers for applications in ISO Class 1 - 3 cleanrooms
  • In ISO Class 4 and above CoverMAX® can lead to significant reductions in both labor and wipe consumption
The top and bottom shell is made of cleanroom grade polyester fabric heat sealed around highly absorbent fine pore foam. This unique construction enables operators to apply large, uniform volumes of sterile alcohol and disinfectants to surfaces that must be decontaminated. The foam core evenly dispenses fluids through the outer polyester shell under normal wiping pressure to enhance fluid coverage and contact time.