Specifications
- Packed:70 wipes/canister
- Description:Disinfectant wipes, blue, in portable canister
- L×W:70×130 mm
- Pk:1
Specifications
About this item
These non woven wipes are made of cellulose and polyester, pre-saturated with a disinfectant solution. The wipes destroy 99,9% of gram-positive and gram-negative bacteria, most viruses, fungi and other micro-organisms.
- Non-tainting result - no rinsing required
- Controlled delivery of disinfectant
- Dermatologically tested
- Portable canister with flip-top lid for easy dispensing
Ideal for cleaning hard surfaces, machinery, utensils and equipment (for example thermometer probes) in potentially sensitive areas including food production, food service, healthcare, general industry and public environments.
Active ingredients: Alkyl Dimethyl Benzyl Ammonium, Chloride/Alkyl, Dimethyl Ethylbenzyl, Ammonium Chloride (Quaternary Ammonium Chlorides) 0,48%; Polymeric Biguanide Hydrochloride 0,2%
Testing: Proven against Clostridium difficile, Clostridium perfringens, Enterococcus Hirae, E.Coli, Listeria, MRSA, TB, Proteus Vulgaris, Pseudomonas Aeruginsoa, Salmonella, Staphylococcus Aureus, VRE, Hepatitis C, SARS, HIV, Influenza A H1N1, Aspergillus Niger and Candida Albicans.