Stereo zoom microscopes, SMZ745 and SMZ745T
: NIKON
These stereo zoom microscopes are suitable for gross observations, regenerative studies, embryology/IVF and environmental science.
- Greenough optics with interpupillary adjustment of 52 to 75 mm
- Working distance of 115 mm for ample manipulation
- Optical path switching lever (SMZ745T only)
Note: Not for sale in Republic of Ireland or Northern Ireland.
: Please contact your local Avantor sales office for further details.
Eyepieces
Frequently Bought Together