Bottles, narrow neck, with standard ground stoppers, BIBBY®
Soda-lime glass, clear or brown, with standard ground joint and polyethylene stopper.
- Good chemical and physical resistance
- For use in applications that are not associated with high thermal loads
- Stable and robust
Available in crystal clear glass or brown tinted to protect light-sensitive substances.
Complies with ISO 4796-2
: Complies with ISO 4796-2
Frequently Bought Together